3-Fluoro-4-methylbenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-Fluoro-4-methylbenzene-1-sulphonyl chloride
L19516.03 L19516.06
L19516.03EA 46.5 EUR
L19516.03 L19516.06
3-Fluoro-4-methylbenzenesulphonyl chloride ≥97%
3-Fluoro-4-methylbenzenesulphonyl chloride
Purity: 97% CAS No.: 90260-13-2 Molecular Formula: C7H6ClFO2S Formula Weight: 208.64 Flash Point: 110°(230°F) Melting Point: 46-50° UN No.: 3261 MDL No.: MFCD03094389
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094389
CAS Number: 90260-13-2
UN: 3261
ADR: 8 II