5-Fluoro-2-methylbenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
5-Fluoro-2-methylbenzene-1-sulphonyl chloride, 3-Fluor-6-methylbenzenesulphonyl chloride
M-Clarity™ quality level = MQ100
558583-1G
558583-5G
SIAL558583-1GEA
52.1
EUR
SIAL558583-1G
SIAL558583-5G
5-Fluoro-2-methylbenzenesulphonyl chloride, Sigma-Aldrich®
5-Fluoro-2-methylbenzenesulphonyl chloride
Formula:
C₇H₆ClFO₂S MW: 208.64 g/mol Boiling Pt: 155…158 °C (50 mmHg) |
MDL Number:
MFCD00052958 CAS Number: 445-05-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...