3-Phenylpropionyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
A10860.09
A10860.30
A10860.09EA
54.6
EUR
A10860.09
A10860.30
3-Phenylpropionyl chloride ≥98%
3-Phenylpropionyl chloride
Purity: 98%
CAS No.: 645-45-4
Molecular Formula: C6H5(CH2)2COCl
Formula Weight: 168.62
Flash Point: 108°(226°F)
Refractive Index: 1.5265
Odour: Acrid
Melting Point: -7°
Boiling Point: 225°
Beilstein Reference: 742586
UN No.: 3265
MDL No.: MFCD00000748
Formula:
C₉H₉ClO MW: 168.62 g/mol Boiling Pt: 225 °C Melting Pt: –7 °C Density: 1.135 Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000748 CAS Number: 645-45-4 EINECS: 211-443-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...