3-Phenylpropionyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
249440-25G
249440-5G
SIAL249440-25GEA
71.5
EUR
SIAL249440-25G
SIAL249440-5G
3-Phenylpropionyl chloride, Sigma-Aldrich®
3-Phenylpropionyl chloride
Formula:
C₉H₉ClO MW: 168.62 g/mol Boiling Pt: 232 °C (1013 hPa) Melting Pt: –2 °C Density: 0.997 g/cm³ (20 °C) |
MDL Number:
MFCD00000748 CAS Number: 645-45-4 EINECS: 211-443-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...