4,16-Dibromo[2.2]paracyclophane ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4,16-Dibromotricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
B25745.03 B25745.14 B25745.06
B25745.03EA 91.5 EUR
B25745.03 B25745.14 B25745.06
4,16-Dibromo[2.2]paracyclophane ≥98%
4,16-Dibromo-[2.2]-paracyclophane
Purity: 98% CAS No.: 96392-77-7 Molecular Formula: C16H14Br2 Formula Weight: 366.10 Melting Point: 249-252° MDL No.: MFCD09953451
Formula: C₁₆H₁₄Br₂
MW: 366.1 g/mol
Melting Pt: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD09953451
CAS Number: 96392-77-7