4,5-Dicyanoimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile
M-Clarity™ quality level = MQ300
554030-1KG
554030-25G
554030-5G
SIAL554030-1KGEA
5110
EUR
SIAL554030-1KG
SIAL554030-25G
SIAL554030-5G
4,5-Dicyanoimidazole, Sigma-Aldrich®
4,5-Dicyanoimidazole
Formula:
C₅H₂N₄ MW: 118.1 g/mol Melting Pt: 176 °C Density: 1.458 g/cm³ (20 °C) |
MDL Number:
MFCD00005194 CAS Number: 1122-28-7 EINECS: 214-344-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...