Luteolin, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone

M-Clarity™ quality level = MQ200

L9283-10MG L9283-50MG
SIALL9283-10MGEA 216 EUR
SIALL9283-10MG SIALL9283-50MG
Luteolin, Sigma-Aldrich®
Luteolin
Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00017309
CAS Number: 491-70-3
EINECS: 207-741-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR