Apigenin

Supplier: Molekula

MOLE22930039-25MGEA 0 EUR
MOLE22930039-25MG
Apigenin
Apigenin
Formula: C₁₅H₁₀O₅
MW: 270,24 g/mol
MDL Number: MFCD00006831
CAS Number: 520-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR