Genistein

Supplier: EHRENSTORFER GMBH

EHERC13999800EA 211 EUR
EHERC13999800
Genistein
Genistein
Formula: C₁₅H₁₀O₅
MW: 270,24 g/mol
Melting Pt: 297…298 °C
MDL Number: MFCD00016952
CAS Number: 446-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR