4,4'-Diaminobenzanilide 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Amino-N-(4-aminophenyl)benzamide
B22419.09 B22419.18
B22419.09EA 32.1 EUR
B22419.09 B22419.18
4,4'-Diaminobenzanilide 98%
4,4'-Diaminobenzanilide
Purity: 98% CAS No.: 785-30-8 Molecular Formula: C13H13N3O Formula Weight: 227.27 Melting Point: 200-202° Beilstein Reference: 2215469
Formula: C₁₃H₁₃N₃O
MW: 227.27 g/mol
Melting Pt: 200…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00025361
CAS Number: 785-30-8
EINECS: 212-321-5