4-Bromo-2,5-difluorobenzenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
632856-1G 632856-5G
SIAL632856-1GEA 31.3 EUR
SIAL632856-1G SIAL632856-5G
4-Bromo-2,5-difluorobenzenesulphonyl chloride, Sigma-Aldrich®
4-Bromo-2,5-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Boiling Pt: 76 °C (0.01 mmHg)
Melting Pt: 34…36 °C
MDL Number: MFCD00042181
CAS Number: 207974-14-9
UN: 3261
ADR: 8,II