4-Bromo-3-methylbenzenesulphonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Bromo-3-methylbenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

556009-1G 556009-5G
SIAL556009-1GEA 65.3 EUR
SIAL556009-1G SIAL556009-5G
4-Bromo-3-methylbenzenesulphonyl chloride, Sigma-Aldrich®
4-Bromo-3-methylbenzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 61 °C
MDL Number: MFCD03094696
CAS Number: 72256-93-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR