4-Chloro-o-toluic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

634034-1G 634034-5G
SIAL634034-1GEA 37.4 EUR
SIAL634034-1G SIAL634034-5G
4-Chloro-o-toluic acid, Sigma-Aldrich®
4-Chloro-o-toluic acid
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00045852
CAS Number: 7499-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR