4-Chloro-3-nitrobenzenesulphonyl chloride

Supplier: Molekula

MOLEM43931765EA 0 EUR
MOLEM43931765
4-Chloro-3-nitrobenzenesulphonyl chloride
4-Chloro-3-nitrobenzenesulphonyl chloride
Formula: C₆H₃Cl₂NO₄S
MW: 256,07 g/mol
Melting Pt: 61,1 °C
MDL Number: MFCD00007440
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR