2-Fluoroisonicotinonitrile, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Cyano-2-fluoropyridine

M-Clarity™ quality level = MQ100

704016-1G 704016-5G
SIAL704016-1GEA 26.1 EUR
SIAL704016-1G SIAL704016-5G
2-Fluoroisonicotinonitrile, Sigma-Aldrich®
2-Fluoroisonicotinonitrile
Formula: C₆H₃FN₂
MW: 122.1 g/mol
MDL Number: MFCD09027299
CAS Number: 3939-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR