4-Fluorobenzenesulphonamide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

542733-1G 542733-5G
SIAL542733-1GEA 121 EUR
SIAL542733-1G SIAL542733-5G
4-Fluorobenzenesulphonamide, Sigma-Aldrich®
4-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
MDL Number: MFCD00025384
CAS Number: 402-46-0
EINECS: 206-946-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR