4-Pentylbenzenesulphonyl chloride
Supplier: Thermo Scientific
Synonyms:
4-Pentylbenzene-1-sulphonyl chloride, 4-Amylbenzenesulphonyl chloride, 4-Amylbenzene-1-sulphonyl chloride
BTB09810DA
BTB09810EA
BTB09810.50
MAYBBTB09810DAEA
0
EUR
MAYBBTB09810DA
MAYBBTB09810EA
MAYBBTB09810.50
4-Pentylbenzenesulphonyl chloride
4-Pentylbenzenesulphonyl chloride
Formula:
C₁₁H₁₅ClO₂S MW: 246.76 g/mol |
MDL Number:
MFCD00173759 CAS Number: 73948-18-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...