(±)-Camphor-10-sulphonic acid, Sigma-Aldrich®

Supplier: Merck

Synonyms: DL-Camphor-10-sulphonic acid, (±)-2-Oxobornane-10-sulphonic acid, (7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid

M-Clarity™ quality level = MQ200

147923-100G 147923-500G
SIAL147923-100GEA 60.4 EUR
SIAL147923-100G SIAL147923-500G
(±)-Camphor-10-sulphonic acid, Sigma-Aldrich®
(±)-Camphor-10-sulphonic acid
Formula: C₁₀H₁₆O₄S
MW: 232.3 g/mol
MDL Number: MFCD00074827
CAS Number: 5872-08-2
EINECS: 227-527-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR