Methyl benzenesulphonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
165956-100G
165956-500G
SIAL165956-100GEA
127
EUR
SIAL165956-100G
SIAL165956-500G
Methyl benzenesulphonate, Sigma-Aldrich®
Methyl benzenesulphonate
Formula:
C₇H₈O₃S MW: 172.2 g/mol Boiling Pt: 75 °C (0.3 mmHg) Melting Pt: 4.5 °C Density: 1.2889 g/cm³ (0 °C) |
MDL Number:
MFCD00014737 CAS Number: 80-18-2 EINECS: 201-256-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...