Methyl benzenesulphonate 99%
Supplier: Thermo Scientific
Danger
413991000 413990250
ACRO413991000EA 71.9 EUR
ACRO413991000 ACRO413990250
Methyl benzenesulphonate 99%
Methyl benzenesulphonate
Formula: C₇H₈O₃S
MW: 172.2 g/mol
Boiling Pt: 280 °C
Melting Pt: –4 °C
Density: 1.3 g/cm³
Flash Pt: 143 °C
Storage Temperature: Ambient
MDL Number: MFCD00014737
CAS Number: 80-18-2
EINECS: 201-256-8
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5180 (20°C, 589 nm)
HPLC ≥98.5 %