Norcamphor, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Oxonorbornane, Bicyclo[2.2.1]heptan-2-one, Bicyclo[2.2.1]-2-heptanone, 2,5-Methanocyclohexanone, 2-Norbornanone

M-Clarity™ quality level = MQ100

N32601-100G N32601-25G N32601-5G
SIALN32601-100GEA 609 EUR
SIALN32601-100G SIALN32601-25G SIALN32601-5G
Norcamphor, Sigma-Aldrich®
Norcamphor
Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 170…173 °C (1013 hPa)
Melting Pt: –88 °C
Density: 1.03 g/cm³ (25 °C)
MDL Number: MFCD00074823
CAS Number: 497-38-1
EINECS: 207-846-1
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR