Bilirubin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-(2-((3-(2-Carboxyethyl)-4-methyl-5-((Z)-(3-methyl-5-oxo-4-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-2-yl)methyl)-4-methyl-5-((Z)-(4-methyl-5-oxo-3-vinyl-1H-pyrrol-2(5H)-ylidene)methyl)-1H-pyrrol-3-yl)propanoic acid
M-Clarity™ quality level = MQ200
B4126-1G
B4126-5G
SIALB4126-1GEA
355
EUR
SIALB4126-1G
SIALB4126-5G
Bilirubin, Sigma-Aldrich®
Bilirubin
Formula:
C₃₃H₃₆N₄O₆ MW: 584.67 g/mol Melting Pt: > 360 °C Density: 1.31 g/cm³ (20 °C) |
MDL Number:
MFCD00005499 CAS Number: 635-65-4 EINECS: 211-239-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...