Diaminomaleonitrile, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
163880-100G
163880-25G
SIAL163880-100GEA
80.3
EUR
SIAL163880-100G
SIAL163880-25G
Diaminomaleonitrile, Sigma-Aldrich®
Diaminomaleonitrile
Formula:
C₄H₄N₄ MW: 108.1 g/mol Boiling Pt: 445 °C (1013 hPa) Melting Pt: 178…181 °C Density: 1.368 g/cm³ (20 °C) |
MDL Number:
MFCD00001870 CAS Number: 1187-42-4 EINECS: 214-697-6 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...