Diaminomaleonitrile 98%
Supplier: Thermo Fisher Scientific
Danger
172700250 172701000 172705000
ACRO172700250EA 34.5 EUR
ACRO172700250 ACRO172701000 ACRO172705000
Diaminomaleonitrile 98%
Diaminomaleonitrile
Formula: C₄H₄N₄
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 178…179 °C
Density: 1,368 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001870
CAS Number: 1187-42-4
EINECS: 214-697-6
UN: 3439
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Brown Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥97.5 %