Coronene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
335355-100MG 335355-500MG
SIAL335355-100MGEA 291 EUR
SIAL335355-100MG SIAL335355-500MG
Coronene, Sigma-Aldrich®
Coronene
Formula: C₂₄H₁₂
Boiling Pt: 525 °C (1013 hPa)
Melting Pt: 437…440 °C
Density: 1.371 g/cm³ (25 °C)
MDL Number: MFCD00004134
CAS Number: 191-07-1
EINECS: 205-881-7