Cerium triacetate hydrate ≥99.995% (metals basis)
Supplier: Thermo Scientific
Danger
Synonyms: Acetic acid cerium (III) salt hydrate, Cerium (III) acetate hydrate
44433.09 44433.18 44433.30
44433.09EA 72.4 EUR
44433.09 44433.18 44433.30
Cerium triacetate hydrate ≥99.995% (metals basis)
Cerium triacetate hydrate
Purity: 99.995% CAS No.: 206996-60-3 Molecular Formula: C6H9CeO6.xH2O Formula Weight: 317.26(anhy) Physical Form: Crystalline Appearance: White MDL No.: MFCD00051060
Formula: C₆H₉CeO₆·H₂O
MW: 335.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150533
CAS Number: 206996-60-3
EINECS: 208-654-0