1-Phenylethyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Styrallyl acetate, α-Methylbenzyl acetate
M-Clarity™ quality level = MQ300
W268402-1KG-K W268402-25KG-K W268402-5KG-K
SIALW268402-1KG-KEA 77.9 EUR
SIALW268402-1KG-K SIALW268402-25KG-K SIALW268402-5KG-K
1-Phenylethyl acetate, Sigma-Aldrich®
1-Phenylethyl acetate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 94 °C (1013 hPa)
Density: 1.058 g/cm³ (16 °C)
MDL Number: MFCD00008858
CAS Number: 93-92-5
EINECS: 202-288-5