2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Diethylene glycol monoethyl ether acetate
M-Clarity™ quality level = MQ200
537527-1L
537527-4L
SIAL537527-1LEA
78.6
EUR
SIAL537527-1L
SIAL537527-4L
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
2-(2-Ethoxyethoxy)ethyl acetate
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 210 Melting Pt: –25 °C Density: 1.0114 g/cm³ (20 °C) |
MDL Number:
MFCD00041928 CAS Number: 112-15-2 EINECS: 203-940-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...