4-Nitrophenyl acetate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

N8130-100G N8130-10G N8130-25G N8130-5G
SIALN8130-100GEA 222 EUR
SIALN8130-100G SIALN8130-10G SIALN8130-25G SIALN8130-5G
4-Nitrophenyl acetate, Sigma-Aldrich®
Formula: C₈H₇NO₄
Melting Pt: 76…79 °C
MDL Number: MFCD00007326
CAS Number: 830-03-5
EINECS: 212-593-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR