18-Pentatriacontanone 85, technical
Supplier: Thermo Fisher Scientific
41445.14 41445.22
41445.14EA 89.3 EUR
41445.14 41445.22
18-Pentatriacontanone 85, technical
18-Pentatriacontanone
Purity: 97+% CAS No.: 504-53-0 Molecular Formula: C35H70O Formula Weight: 506.94 Physical Form: Powder Appearance: Off-white Melting Point: 79-80° Solubility: Soluble in benzene MDL No.: MFCD00048491
Antiblocking agent
Formula: C₃₅H₇₀O
MW: 506.94 g/mol
Boiling Pt: 543
Melting Pt: 79…80 °C
Density: 0.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00048491
CAS Number: 504-53-0
EINECS: 207-993-1