Potassium ethanolate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Potassium ethylate, Potassium ethoxide
M-Clarity™ quality level = MQ200
333379-100G
333379-500G
SIAL333379-100GEA
80.2
EUR
SIAL333379-100G
SIAL333379-500G
Potassium ethanolate, Sigma-Aldrich®
Potassium ethanolate
Formula:
C₂H₅KO MW: 84.16 g/mol |
MDL Number:
MFCD00054277 CAS Number: 917-58-8 EINECS: 213-029-0 UN: 3206 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...