Methoxyacetic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

194557-250G 194557-50G
SIAL194557-250GEA 97.8 EUR
SIAL194557-250G SIAL194557-50G
Methoxyacetic acid, Sigma-Aldrich®
Methoxyacetic acid
Formula: C₃H₆O₃
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.18 g/cm³ (20 °C)
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR