Methoxyacetic acid 97+%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 2-Methoxyacetic acid
186770010 186770250 186772500
ACRO186770010EA 492 EUR
ACRO186770010 ACRO186770250 ACRO186772500
Methoxyacetic acid 97+%
Methoxyacetic acid
Formula: C₃H₆O₃
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.174 g/cm³
Flash Pt: 121 °C
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
Titration with NaOH 97.0 to 103.0 %
HPLC ≥97.5 %