Sodium ethoxide 96%, pure
Supplier: Thermo Scientific
Danger
Synonyms: Sodium ethylate, Sodium ethanolate
168592500 168595000 168590025
ACRO168592500EA 45.1 EUR
ACRO168592500 ACRO168595000 ACRO168590025
Sodium ethoxide 96%, pure
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260 °C (dec.)
Density: 0,868 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3095
ADR: 8,I
Order Now
Specification Test Results
Appearance White to yellow Powder
Infrared spectrum Conforms
Assay ≥95 % (NaOC2H5)
Titration with HCl ≥98 % (total NaOC2H5)
NaOH + Na2CO3 (as NaOH) ≤2 %