Sodium ethoxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium ethylate, Sodium ethanolate
M-Clarity™ quality level = MQ100
71210-250G 71210-50G
SIAL71210-250GEA 84.6 EUR
SIAL71210-250G SIAL71210-50G
Sodium ethoxide, Sigma-Aldrich®
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Melting Pt: 260 °C
Density: 0.868 g/cm³ (20 °C)
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 3206
ADR: 4.2,II