Isobutyl phenylacetate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W221007-10KG-K W221007-1KG-K
SIALW221007-10KG-KEA 549 EUR
SIALW221007-10KG-K SIALW221007-1KG-K
Isobutyl phenylacetate, Sigma-Aldrich®
Isobutyl phenylacetate
Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD00026482
CAS Number: 102-13-6
EINECS: 203-007-9