Acrylic acid stabilised, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Prop-2-enoic acid

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

147230-100G 147230-18KG 147230-1L 147230-2.5L 147230-3KG 147230-500G 147230-5G
SIAL147230-100GEA 49.8 EUR
SIAL147230-100G SIAL147230-18KG SIAL147230-1L SIAL147230-2.5L SIAL147230-3KG SIAL147230-500G SIAL147230-5G
Acrylic acid stabilised, Sigma-Aldrich®
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 141.6
Melting Pt: 13
Density: 1.06 g/cm³ (10 °C)
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR