trans-2-Methyl-2-butenal, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal
M-Clarity™ quality level = MQ100
192619-25G
192619-5G
SIAL192619-25GEA
92.2
EUR
SIAL192619-25G
SIAL192619-5G
trans-2-Methyl-2-butenal, Sigma-Aldrich®
trans-2-Methyl-2-butenal
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 115…117 °C (1013 hPa) Density: 0.8688 g/cm³ (20 °C) |
MDL Number:
MFCD00006977 CAS Number: 497-03-0 EINECS: 207-833-0 UN: 1989 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...