trans-4-Aminocyclohexanol 97%
Supplier: Thermo Scientific
Danger
346680050 346680250 346681000
ACRO346680050EA 34.8 EUR
ACRO346680050 ACRO346680250 ACRO346681000
trans-4-Aminocyclohexanol 97%
trans-4-Aminocyclohexanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 127 °C (14 mmHg)
Melting Pt: 108…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00067698
CAS Number: 27489-62-9
EINECS: 248-492-8
UN: 3259
ADR: 8,III
Order Now
Specification Test Results
Appearance White to cream to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 108 to 113 °C
GC ≥96.0 %