2-Tridecanone ≥98%
Supplier: Thermo Scientific
A19042.14
A19042.22
A19042.14EA
37.2
EUR
A19042.14
A19042.22
2-Tridecanone ≥98%
2-Tridecanone
Purity: 98+%
CAS No.: 593-08-8
Molecular Formula: C13H26O
Formula Weight: 198.35
Flash Point: 122°(251°F)
Refractive Index: 1.4350
Odour: Characteristic
Melting Point: 29-31°
Boiling Point: 133-134°/10mm
Beilstein Reference: 1757402
UN No.: 3082
MDL No.: MFCD00008968
Formula:
C₁₃H₂₆O MW: 198.35 g/mol Boiling Pt: 133…134 °C (10 mmHg) Melting Pt: 29…31 °C Density: 0.822 Flash Pt: 122 °C (251 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008968 CAS Number: 593-08-8 EINECS: 209-784-0 UN: 3077 ADR: 9,III |