4-Heptanone

Supplier: EHRENSTORFER GMBH

EHERC14127800EA 47.2 EUR
EHERC14127800
4-Heptanone
4-Heptanone
Formula: C₇H₁₄O
MW: 114,19 g/mol
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –34 °C
Density: 0,8116 g/cm³ (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00009403
UN: 2710
ADR: 3,III
Merck Index: 13,03379

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR