Petri dish racks, Nalgene®
Supplier: Thermo Fisher Scientific
NALG5922-0024EA 110 EUR
NALG5922-0024
Petri dish racks, Nalgene®
Racks Petri Dish Racks
Petri dish rack with acrylic frame and PC posts.
  • Lightweight
  • Easy to load
  • Provides safe transport
Break-resistant PC posts. Supplied unassembled. Holds up to twenty-four 100 mm dishes.