Ethyl acetate ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 99.5+%
CAS No.: 141-78-6
Molecular Formula: C4H8O2
Formula Weight: 88.11
Flash Point: -3°(26°F)
Physical Form: Liquid
Density: 0.902
Refractive Index: 1.3720
Appearance: Colourless
Odour: Sweet
Melting Point: -84°
Boiling Point: 77°
Beilstein Reference: 506104
Merck Reference: 14,3757
UN No.: 1173
MDL No.: MFCD00009171
Formula:
CH₃COOC₂H₅ Boiling Pt: 77 °C Melting Pt: –84 °C Density: 0.902 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 14,3757 |