Amyl acetate, Sigma-Aldrich®

Supplier: Merck

SIAL46022-1L-FEA 119 EUR
SIAL46022-1L-F SIAL46022-250ML-F
Amyl acetate, Sigma-Aldrich®
Amyl acetate
Formula: C₇H₁₄O₂
MW: 130,19 g/mol
Boiling Pt: 149 °C (1013 hPa)
Melting Pt: –71 °C
Density: 0,876 g/cm³ (20 °C)
Flash Pt: 23 °C
MDL Number: MFCD00009500
UN: 1104
ADR: 3,III
https://si.vwr.com/stibo/web/std.lang.all/27/75/722775.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR