Palmitic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: Hexadecanoic acid, Hexadecylic acid
MQ200
27734-25KG 27734-1KG
SIAL27734-25KGEA 747 EUR
SIAL27734-25KG SIAL27734-1KG
Palmitic acid, Sigma-Aldrich®
Palmitic acid
Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 352
Melting Pt: 61…63 °C
Density: 0.853 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002747
CAS Number: 57-10-3
EINECS: 200-312-9
Merck Index: 13,07064