Formic acid
Supplier: Roth Carl
Danger
4724.1 4724.2
ROTH4724.1EA 58.7 EUR
ROTH4724.1 ROTH4724.2
Formic acid
Formic acid
Formula: HCO₂H
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8.2…8.4 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM