Lignoceric acid 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Tetracosanoic acid
215640010 215640050 215642500
ACRO215640010EA 160 EUR
ACRO215640010 ACRO215640050 ACRO215642500
Lignoceric acid 99%
Lignoceric acid
Formula: C₂₄H₄₈O₂
MW: 368.64 g/mol
Boiling Pt: 272 °C (10 mmHg)
Melting Pt: 84…86 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002810
CAS Number: 557-59-5
EINECS: 209-180-7
Merck Index: 13,05509
Order Now
Specification Test Results
Appearance White Powder or crystals or flakes
Infrared spectrum Conforms
GC ≥98.5 %