Tridecanoic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

T0502-5G T0502-10G T0502-1G
SIALT0502-5GEA 51.7 EUR
SIALT0502-5G SIALT0502-10G SIALT0502-1G
Tridecanoic acid, Sigma-Aldrich®
Tridecanoic acid
Formula: C₁₃H₂₆O₂
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 41.5 °C
Density: 1.025 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00002741
CAS Number: 638-53-9
EINECS: 211-341-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR