Potassium acetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Acetic acid potassium salt
60035-1KG 60035-250G 60035-50G
SIAL60035-1KGEA 211 EUR
SIAL60035-1KG SIAL60035-250G SIAL60035-50G
Potassium acetate, Sigma-Aldrich®
Potassium acetate
Formula: CH₃COOK
Melting Pt: 304 °C
Density: 1.57 g/cm³ (25 °C)
Flash Pt: >250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM