Sodium propionate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Propionic acid sodium salt, Sodiumpropionate

M-Clarity™ quality level = MQ200

18108-1KG 18108-25KG
SIAL18108-1KGEA 64.8 EUR
SIAL18108-1KG SIAL18108-25KG
Sodium propionate, Sigma-Aldrich®
Sodium propionate
Formula: C₃H₅NaO₂
MW: 96.06 g/mol
Melting Pt: 288.76 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002759
CAS Number: 137-40-6
EINECS: 205-290-4
Merck Index: 12,08816

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR